Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho-Akt (Tyr450), Polyclonal, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 071643/DEL

Catalog No. 50-172-299

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »


Description

Description

Akt (protein kinase B), a serine/threonine kinase, has emerged as a critical enzyme in signal transduction pathways involved in cell proliferation, apoptosis, angiogenesis, and diabetes. In mammals three isoforms of Akt (α, β, γ or Akt 1, 2, 3) are reported that exhibit a high degree of homology, but differ slightly in the localization of their regulatory phosphorylation sites. Akt α is the predominant isoform in most tissues, whereas the highest expression of Akt β is observed in the insulin-responsive tissues, and Akt γ is abundant in brain tissue. Each Akt isoform is composed of three functionally distinct regions: an N-terminal pleckstrin homology (PH) domain that provides a lipid-binding module to direct Akt to PIP2 and PIP3, a central catalytic domain, and a C-terminal hydrophobic motif.

Host Species: Rabbit
Cross Reactivity: Bovine, Chicken, Equine, Human, Mouse, Rat
Immunogen: KLH-conjugated linear peptide corresponding to human phosphorylated AKT at and around Tyr450.

Western Blotting

Specifications

Specifications

phospho-Akt (Tyr450)
Affinity Purified
KLH-conjugated linear peptide corresponding to human phosphorylated AKT at and around Tyr450.
Primary
Bovine, Chicken, Equine, Human, Murine, Rat
Unconjugated
Rabbit
100μg
Polyclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit