Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Akt1/PKBα (Ser473) Rabbit anti-Human, Mouse, Clone: SK703, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04736
Description
Anti-Akt1 Antibody, clone SK703 detects level of phospho-Akt1/PKBα (Ser473) and has been published and validated for use in western blotting.Specifications
phospho-Akt1/PKBα (Ser473) | |
Monoclonal | |
Unconjugated | |
NM_005163 | |
Peptide containing a pSer that corresponds to amino acid position 473 of human Akt1/PKBa | |
Primary | |
Human, Mouse | |
Supernatant |
Western Blot | |
SK703 | |
Cultured supernantant in 0.05% sodium azide | |
Rabbit | |
100 μL | |
AKT1, RAC, PRKBA, RAC-ALPHA, PKB, MGC99656, RAC-PK-alpha, C-AKT, AKT, EC 2.7.11.1 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only