Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Blooms Syndrome (Thr99) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA523026

Catalog No. PIPA523026


Add to cart

Description

Description

Blooms Syndrome Protein Blm is a member of the RecQ helicase protein family. The protein contains 1417 amino acids; including one ATP binding site, one DEAH box, and two putative nuclear localization signals, as well as phosphorylation sites. Blooms Syndrome Protein Blm is known to associate and be phosphorylated by the ATR (ataxia telangiectasia, rad3+) protein, in response to genetic stress. This gene product is thought to play a role in the etiology of two major human health problems, each of which is a very common complication of Bloom ft.s syndrome, namely cancer and diabetes.
Specifications

Specifications

Phospho-Blooms Syndrome (Thr99)
Polyclonal
Unconjugated
BLM
Liquid
BLM
Synthetic phospho-peptide surrounding the Blooms Syndrome Protein Blm threonine 99 region
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Immunocytochemistry, Immunofluorescence, Western Blot
4.8 mg/mL
tris HCl with 0.2M glycine and 0.1% sodium azide; pH 8.5
P54132
BS, RECQ2, RECQL2, RECQL3, Bloom syndrome protein, DNA helicase, RecQ-like type 2, recQ protein-like 3
Rabbit
IgG
50 μL
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
641
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit