Learn More
Phospho-c-Met (Tyr1349) Rabbit anti-Human, Polyclonal, Invitrogen™ Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen 44892G
Catalog No. 44-892-G
Add to cart
Description
Description
The c-Met oncogene was originally isolated from a chemical carcinogen-treated human osteogenic sarcoma cell line by transfection analysis in NIH/3T3 cells. The Met proto-oncogene product was identified as a trans-membrane receptor-like protein with tyrosine kinase activity that is expressed in many tissues. The c-Met gene product has been identified as the cell surface receptor for hepatocyte growth factor, a plasminogen-like protein thought to be a humoral mediator of liver regeneration.
Specifications
Specifications
Phospho-c-Met (Tyr1349)
Polyclonal
Dulbecco′s PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P08581
EC 2.7.10.1, HGF receptor, HGF-SF receptor, Hepatocyte growth factor receptor precursor, Met proto- oncogene tyrosine kinase, c-met, kinase EC 2.7.10.1, HGF receptor, HGF-SF receptor, Hepatocyte growth factor receptor precursor, Met proto- oncogene tyros
Rabbit
IgG
100 μL
-20°C
Primary
4233
Western Blot
Unconjugated
MET
Liquid
MET
The antiserum was produced against a chemically synthesized phosphopeptide derived from the region of human c-Met that contains tyrosine 1349. The sequence is conserved in mouse and rat.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human