Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Caveolin 2 (Ser23) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA125625

Catalog No. PA125625


Add to cart

Description

Description

PA1-25625 detects Caveolin 2 (phospho S23) from human samples. PA1-25625 has been successfully used in Western blot procedures. The PA1-25625 immunogen is a synthetic peptide corresponding to residues 18-31 of human caveolin 2, phosphorylated on serine 23.

The protein encoded by this gene is a major component of the inner surface of caveolae, small invaginations of the plasma membrane, and is involved in essential cellular functions, including signal transduction, lipid metabolism, cellular growth control and apoptosis. This protein may function as a tumor suppressor. CAV1 and CAV2 are located next to each other on chromosome 7 and express colocalizing proteins that form a stable hetero-oligomeric complex. Two transcript variants encoding distinct isoforms have been identified for this gene. By using alternative initiation codons in the same reading frame, two isoforms (alpha and beta) are encoded by one transcript.
Specifications

Specifications

Phospho-Caveolin 2 (Ser23)
Polyclonal
Unconjugated
CAV2
Liquid
CAV2
Synthetic peptide corresponding to residues 18-31 of human caveolin 2, phosphorylated on serine 23.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
0.5 mg/mL
antibody with 1% BSA, 50% glycerol and 0.01% thimerosal
P51636
CAV, MGC12294
Rabbit
IgG
25 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
858
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit