Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-Cortactin (Tyr421), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126704

Catalog No. PA126704

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26704 detects Cortactin (phospho Y421) from mouse samples. PA1-26704 will not react with human.PA1-26704 has been successfully used in Western blot procedures.The PA1-26704 immunogen is a synthetic phosphopeptide corresponding to a portion of mouse Cortactin that contains tyrosine 421.
Specifications

Specifications

Phospho-Cortactin pTyr421
Polyclonal
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Q60598
1110020L01Rik, Ems1
Rabbit
IgG
50μL
Store at -20°C, Avoid Freeze/Thaw Cycles
Primary
13043
Western Blot
Unconjugated
Cttn
Purified
CTTN
Synthetic phosphopeptide corresponding to a portion of mouse Cortactin that contains tyrosine 421.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit