Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-CREB (Ser129, Ser133), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA124754

Catalog No. PA124754

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a transcription factor that is a member of the leucine zipper family of DNA binding proteins. This protein binds as a homodimer to the cAMP-responsive element, an octameric palindrome. The protein is phosphorylated by several protein kinases, and induces transcription of genes in response to hormonal stimulation of the cAMP pathway. Alternate splicing of this gene results in two transcript variants encoding different isoforms.

PA1-24754 detects CREB (phospho S129 + S133) from human and mouse samples. PA1-24754 is expected to cross react with rat (100% conserved) due to sequence homology.PA1-24754 has been successfully used in Western blot procedures.The PA1-24754 immunogen is a synthetic phosphopeptide corresponding to a portion of human CREB containing serines 129 and 133.
Specifications

Specifications

Phospho-CREB pSer129+133
Polyclonal
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P16220, Q01147, P15337
CREB, MGC9284
Rabbit
IgG
50μL
Avoid Freeze/Thaw Cycles
Primary
1385
Western Blot
Unconjugated
CREB1
Purified
CREB1
Synthetic phosphopeptide corresponding to a portion of human CREB containing serines 129 and 133.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit