Learn More
Phospho-Dynamin 1 (Ser778) Sheep anti-Rat, Polyclonal, Invitrogen™
Sheep Polyclonal Antibody
Manufacturer:  Invitrogen PA14621
Catalog No. PA14621
Add to cart
Description
Description
This antibody is predicted to react with bovine, canine, chicken, human and mouse based on 100% sequence homology. This antibody is specific for the ~95 kDa Dynamin protein phosphorylated at Ser778 in Western blots of rat brain lysates.
Dynamin is a member of a group of nerve terminal proteins called dephosphins that regulate synaptic vesicle endocytosis Cousin et al., 2001; Graham et al., 2002; Tsuboi et al., 2002). Cyclin dependent protein kinase 5 phosphorylates dynamin at Ser774 and Ser778 that are the phosphorylation sites on dynamin phosphorylated in vivo (Tan et al., 2003). Phosphorylation of these sites on dynamin is thought to play a key role in synaptic vesicle trafficking.
Specifications
Specifications
Phospho-Dynamin 1 (Ser778)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P21575
DNM, DNM1, Dynamin1, Neuronal dynamin
Sheep
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
140694
Western Blot
Unconjugated
Dnm1
Liquid
DNM1
Synthetic phospho-peptide corresponding to amino acid residues surrounding Ser778 of rat DNM1 conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Rat