Learn More
Phospho-EGFR (Tyr1173) Mouse anti-Human, Clone: 3G3.2, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 051004
Catalog No. 05-100-4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect phospho-EGFR (Tyr1173) using this Anti-phospho-EGFR (Tyr1173) Antibody, clone 3G3.2 validated for use in western blot, flow cytometry, immunohistochemistry and immunocytochemistry.
Specifications
Specifications
phospho-EGFR (Tyr1173)
3G3.2
Purified rabbit ascites containing 0.1MTris-glycine, pH 7.4, 150mM NaCl and 0.05% sodium azide.
Purified
Murine
IgG1, κ, κ
100μg
Primary
Human
Flow Cytometry, Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
NP_005219.2
C14orf27, CEG1, EGFR-5, EGFR5, OTTHUMP00000178873
KLH-conjugated linear peptide corresponding to residues surrounding Tyr1173 of EGFR.
Protein G purfied
2-8°C for up to 12 months.
Monoclonal