Learn More
MilliporeSigma™ anti-phospho-EGFR (Tyr1173), ™ (Upstate™) Non-distribution product as customer accommodation.
Manufacturer:  MilliporeSigma™ 04341/DEL
Catalog No. 50-171-341
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
GenBank Number: NP_005219. MW: 170kDa.
Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting
Abbreviations: AP=Affinity Purification, ChIP=Chromatin IP, CM=Confocal Microscopy, DB=Dot Blot, EIA=ELISA, EM=Electron Microscopy, FC=Flow Cytometry, GS=Gel Supershift, IC=Immunocytochemistry, IF=Immunofluorescence, IH=Immunohistology/Immunohistochemistry, IP=Immunoprecipitation, IPK=IP Kinase Assay, KA=Kinase Assay, NEUT=Neutralization, PIA=Peptide Inhibition Assay, RIA=Radioimmunossay, TCA=T Cell Activation, WB=Western Blotting, Species: Am=Amphibian, Av=Avian, B=Bovine, Ca=Cat ,Ck=Chicken, D=Dog, Dr=Deer, Ds=Drosophila, El=Elk, Fg=Frog, Fs=Fish, Ft=Ferret, G=Goat, Gp=Guinea Pig, H=Horse, Hm=Hamster, Hu=Human, K=Kangaroo, Ms=Mouse, Nhp=Non-Human Primate, O=Ovine, P=Porcine, Rb=Rabbit, Rp=Reptile, Rt=Rat, Ys=Yeast, Xn=Xenopus, (wk)=cross reacts weakly
Specifications
Specifications
phospho-EGFR (Tyr1173)
Semi-purified
KLH-conjugated synthetic peptide corresponding to the amino acid residues including and surrounding tyrosine 1173 of the human EGF Receptor (EGFR)
100μL
Cell Signaling
Monoclonal
Unconjugated
Rabbit
IgG
RUO
Primary
Human