Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-EGFR (Tyr845) Mouse anti-Human, Mouse, Clone: 12A3, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 04283
Description
Detect phospho-EGFR (Tyr845) using this Anti-phospho-EGFR (Tyr845) Antibody, clone 12A3 validated for use in ELISA, immunocytochemistry, immunoprecipitation and western blotting.Specifications
phospho-EGFR (Tyr845) | |
Monoclonal | |
Unconjugated | |
NM_005228.3; NM_201282.1; NM_201283.1; NM_201284.1 | |
Mouse | |
Subsequent thiophilic adsorption and size exclusion chromatography | |
Primary | |
−20°C for 24 months. | |
IgG1 |
ELISA, Immunocytochemistry, Immunoprecipitation, Western Blot | |
12A3 | |
100μg of mouse monoclonal IgG1 lyophilized in 2X PBS containing 0.09% sodium azide, PEG, and sucrose | |
EGFR, ERBB1, ERBB, mENA | |
KLH-conjugated synthetic peptide encompassing the surrounding amino acids of Tyr 845 in human EGFR. | |
100 μg | |
Human, Mouse | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only