Learn More
Phospho-EphB2 (Tyr317) Rabbit anti-Chicken, Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14623
Catalog No. PA14623
Add to cart
Description
Description
This antibody is predicted to react with bovine, human, mouse, Xenopus and zebrafish based on 100% sequence homology. This antibody is specific for the ~46 Dak EphrinB protein phosphorylated at Tyr317. Immunolabeling can be blocked by lambda-phosphatase treatment.
Ephrin receptors and their ligands, the ephrins, mediate numerous developmental processes, particularly in the nervous system. Based on their structures and sequence relationships, ephrins are divided into the ephrin-A (EFNA) class, which are anchored to the membrane by a glycosylphosphatidylinositol linkage, and the ephrin-B (EFNB) class, which are transmembrane proteins. The Eph family of receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. Ephrin receptors make up the largest subgroup of the receptor tyrosine kinase (RTK) family. EphB4 binds to ephrin-B2 and plays an essential role in vascular development.
Specifications
Specifications
Phospho-EphB2 (Tyr317)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P28693
EFL6, EFNB3, EPH related receptor transmembrane ligand ELK L3, EPH related receptor tyrosine kinase ligand 8, Ephrin B3, EPLG8, LERK8
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
313633, 396513
Western Blot
Unconjugated
Ephb2
Liquid
EPHB2
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr317 of chicken EPHB2 conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Chicken, Rat