Learn More
Phospho-Erk1/2 (Thr202/Tyr204, Thr185/Tyr187) Rabbit anti-Human, Mouse, Rat, Clone: AW39R, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05797R
Catalog No. 05-797-RMI
Add to cart
Description
Description
Anti-phospho-Erk1/2 (Thr202/Tyr204 Antibody, Thr185/Tyr187), recombinant clone AW39R detects level of phospho-Erk1/2 (Thr202/Tyr204 and has been published and validated for use in Mplex and western blotting.
Specifications
Specifications
phospho-Erk1/2 (Thr202/Tyr204
AW39R
Protein A Purified immunoglobulin in 30% glycerol, 0.07M Tris-glycine, pH 7.4, 0.105M NaCl, 0.035% sodium azide as a preservative.
Purified
Phosphorylated peptide corresponding to amino acids surrounding the pTEpY motif in the activation loop of human phospho-Erk1/2, in which the Thr and Tyr residues are phosphorylated
Protein A purfied
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human, Murine, Rat
Immunoassay, Multiplexing, Western Blot
Unconjugated
NP_002736, NP_002737
Rabbit
IgG
100μL
Primary
Erk1, Erk2, p44-MAPK, p42-MAPK, MAPK2, PRKM2, p41mapk, PRKM1, p44, ERK2, P42MAPK, ERK, p42, MAPK1, ERT1, ERT2, Mitogen-activated protein kinase, MAP Kinase 2, MAP Kianse 1, MAPK