Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Estrogen Receptor alpha (Ser118) Rabbit anti-Human, Clone: NL44, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 05793
Description
Anti-phospho-Estrogen Receptor alpha (Ser118) Antibody, clone NL44 is a Rabbit Monoclonal Antibody for detection of ER alpha also known as Estrogen receptor alpha and has been tested in western blotting.Specifications
phospho-Estrogen Receptor alpha (Ser118) | |
Monoclonal | |
Unconjugated | |
NM_000125 | |
Peptide containing the sequence QLpSPF in which pS corresponds to phosphoserine at residue 118 of human estrogen receptor alpha | |
100 μL | |
ESR1, Era, ER-alpha, ESR, ER, NR3A1, ESRA, DKFZp686N23123 | |
−20°C for 24 months | |
IgG |
Western Blot | |
NL44 | |
0.2M Tris-glycine, pH 7.4, 0.15M NaCl, 100mM EDTA, 0.05% sodium azide, before the addition of glycerol to 30% | |
Rabbit | |
Protein A purfied | |
Primary | |
Human | |
Purified |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction