Learn More
MilliporeSigma™ anti-phospho-Estrogen Receptor alpha (Ser305), Clone: 124.9.4, Rabbit Monoclonal,
Manufacturer:  MilliporeSigma™ 05922R
Catalog No. 05-922-RMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
phospho-Estrogen Receptor alpha (Ser305)
124.9.4
Purified Rabbit Monoclonal IgG in buffer containing PBS with 0.05% sodium azide.
Purified
KLH-conjugated linear peptide corresponding to human Estrogen Receptor alpha phosphorylated at Ser305.
Protein A purified
2-8°C for 12 months
Monoclonal
Human
Immunohistochemistry, Western Blot
Unconjugated
NP_000116
Rabbit
IgG
100μg
Primary
ESR1, ESR, NR3A1