Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phospho-FAK/PTK2 pTyr576 Clone: 2H74L24, Invitrogen™

Phospho-FAK/PTK2 pTyr576 Monoclonal antibody 700013 from Thermo Scientific™ Pierce™ specifically detects Phospho-FAK/PTK2 pTyr576 in Human samples, and it is validated for IF and ICC

Manufacturer:  Invitrogen 700013

Encompass

Catalog No. 70-001-3


Add to cart

Description

Description

Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of integrin signaling. FAK plays a central role in cell spreading, differentiation, migration, cell death and acceleration of the G1 to S phase transition of the cell cycle. Regulation of FAK includes phosphorylation at multiple tyrosine and serine residues. Increased FAK tyrosine phosphorylation occurs upon integrin engagement with fibronectin.

700013 detects Phospho-FAK/PTK2 pTyr576 in Human samples. 700013 has been successfully used in IF and ICC procedures. The immunogen for 700013 is a peptide corresponding to amino acids 573-582 of Q05397.

Applications:

Immunocytochemistry, Immunofluorescence

Specifications

Specifications

Phospho-FAK/PTK2 pTyr576
Unconjugated
Q05397
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
2H74L24
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
A peptide corresponding to amino acids 573-582 of Q05397.
Protein A
Primary
5747
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit