Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-GluR1 (Ser831) Rabbit anti-Chicken, Human, Mouse, Rat, Clone: N453, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04823
Description
Anti-phospho-GluR1 (Ser831) Antibody, clone N453 detects level of phospho-GluR1 (Ser831) and has been published and validated for use in western blotting.Specifications
phospho-GluR1 (Ser831) | |
Monoclonal | |
Unconjugated | |
NP_000818 | |
KLH-conjugated, synthetic peptide containing the sequence ...QQ[pS]IN... in which pS corresponds to phospho-serine at position 831 of the GluR1 mature chain. Sequence is identical in human, rat, mouse and chicken. | |
Primary | |
Chicken, Human, Mouse, Rat | |
Supernatant |
Western Blot | |
N453 | |
Cultured supernantant in 0.05% sodium azide. | |
Rabbit | |
100 μL | |
GRIA1, GluR-A, GluR-1, GLUR1, GluR-K1, GLURA, HBGR1, GLUH1, MGC133252 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only