Learn More
Phospho-GSK3 (Tyr279/Tyr216) Mouse anti-Human, Mouse, Rat, Clone: 5G-2F, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05413
Catalog No. 05-413-MI
Add to cart
Description
Description
This Anti-phospho-GSK3 (Tyr279/Tyr216) Antibody, clone 5G-2F is validated for use in western blotting for the detection of phospho-GSK3 (Tyr279/Tyr216).
Specifications
Specifications
phospho-GSK3 (Tyr279/Tyr216)
5G-2F
Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-glycine, pH 7.4, 0.15MNaCl, 0.05% sodium azide. Frozen at −20deg;C.
Purified
Murine
κ
200μg
Primary
Human, Murine, Rat
Western Blot
Unconjugated
NP_476715.1
gskt, CG31003
Synthetic peptide (C-KQLLHGEPNVS[pY]ICSRY) corresponding to amino acids 203-219 derived from a region of the catalytic domain (between subdomain VII and VIII) of the Drosophila GSK3/shaggy enzyme coupled to ovalbumin.
Protein G chromatography
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles.
Monoclonal