Learn More
Phospho-Histone H1 (Thr165) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061370
Catalog No. 06-137-0MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Use Anti-phospho-Histone H1 (Thr165) Antibody (Rabbit Polyclonal Antibody) validated in western blot, DB to detect phospho-Histone H1 (Thr165) also known as Histone H1.2, Histone H1d.
Specifications
Specifications
phospho-Histone H1 (Thr165)
Unconjugated
Affinity Purified
Rabbit
Antigen Affinity purified
2-8°C for 12 months
Polyclonal
Human
Dot Blot, Western Blot
Purified rabbit polyclonal serum in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
HIST1H1C; H1F2
KLH-conjugated linear peptide corresponding to human Histone H1 phosphorylated at Thr165.
100μg
Primary
NP_005310