Learn More
Phospho-Histone H3 (Ser10) Rabbit anti-Human, Clone: 63-1C-8, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05817RI
Catalog No. 05-817-RIMI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-phospho-Histone H3 (Ser10) Antibody, clone 63-1C-8, is a recombinant rabbit monoclonal antibody that detects Histone H3 phosphorylated at serine 10. This purified recombinant mAb, also known as Anti-H3S10p, is published and validated in western blot, Flow Cytometry, and is specificity verified by DB.
Specifications
Specifications
phospho-Histone H3 (Ser10)
63-1C-8
Purified rabbit monoclonal IgGκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Purified
Rabbit
IgG, κ, κ
100μL
Primary
Human
Flow Cytometry, Western Blot
Unconjugated
NP_003484
HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886
KLH-conjugated linear peptide corresponding to Human Histone H3 phosphorylated at Ser10.
Protein A purified
2-8°C for one year
Monoclonal