Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-Histone H3 (Ser10) Rabbit anti-Human, Clone: MC463, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04817
Description
Anti-phospho-Histone H3 (Ser10) Antibody, clone MC463 is a rabbit monoclonal antibody for detection of Histone H3 phosphorylated at serine 10. This mAb, also known as H3S10p, is specificity verified using DB and Mplex, published in peer reviewed journals and validated in Chimmunoprecipitation, immunofluoresence, western blot, immunocytochemistry, ChIP-seq.Specifications
phospho-Histone H3 (Ser10) | |
Monoclonal | |
Unconjugated | |
NM_003493 | |
Peptide containing the sequence RK[pS]TG. in which pS corresponds to phospho-serine at position 10 of human histone H3 | |
Primary | |
Human | |
Supernatant |
Multiplex, Dot Blot, Immunocytochemistry, Immunofluorescence, Immunoassay, Western Blot | |
MC463 | |
Cultured supernantant in 0.05% sodium azide. | |
Rabbit | |
100 μL | |
HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ, HIST3H3, H3FT, H3T, MGC126886 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only