Learn More
Phospho-Histone H3 (Thr3) Rabbit anti-Human, Clone: JY325, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05746R
Catalog No. 05-746-RMI
Add to cart
Description
Description
Anti-phospho-Histone H3 (Thr3) Antibody, clone JY325 is a Rabbit Monoclonal Antibody for detection of phospho-Histone H3 (Thr3) also known as Histone H3.1 and has been validated in western blot, PIA.
Specifications
Specifications
phospho-Histone H3 (Thr3)
JY325
Purified Rabbit Monoclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide and 40% glycerol.
Purified
KLH-conjugated linear peptide corresponding to human Histone H3 phosphorylated at Thr3.
Protein A
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles.
Monoclonal
Human
Inhibition Assays, Western Blot
Unconjugated
NP_066298
Rabbit
IgG
100μL
Primary
HIST1H3A, H3FA, HIST1H3B, H3FL, HIST1H3C, H3FC, HIST1H3D, H3FB, HIST1H3E, H3FD, HIST1H3F, H3FI, HIST1H3G, H3FH, HIST1H3H, H3FK, HIST1H3I, H3FF, HIST1H3J, H3FJ
Documents
Promotions
Promotions
  • MilliporeSigma
    Buy two qualifying MilliporeSigma antibodies and get a third of equal or lesser value at no additional cost.
    12-31-2020