Learn More
MilliporeSigma™ anti-phospho-IGF-IR (Tyr1131)/InsR (Tyr1158), Clone: JY202,
Manufacturer:  MilliporeSigma™ 05694
Catalog No. 05-694-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
phospho-IGF-IR (Tyr1131)/InsR (Tyr1158)
JY202
Protein G Purified immunoglobulin in 0.1M Tris-glycine, pH 7.4, 0.15M NaCl with 0.05% sodium azide as a preservative, before the addition of 30% glycerol.
Purified
Murine
IgG2a, κ, κ
200μg
Primary
Human, Murine
Immunoprecipitation, Western Blot
Unconjugated
A25690
IGF1R, IGFIR, MGC18216, MGC142170, JTK13, MGC142172, CD221
peptide containing the amino acids (DI[pY]ET) in which pY corresponds to tyrosine 1131 of human IGF-1 receptor and tyrosine 1158 (mature protein; 1185 of the precursor protein) of human Insulin Receptor
Protein G purfied
−20°C in undiluted aliquots for up to 24 months. Avoid repeated freeze/thaw cycles.
Monoclonal