Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-IGF1R beta (Tyr1158, Tyr1162, Tyr1163), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126725

Catalog No. PIPA126725

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26725 detects Insulin Receptor / IGF1R (phospho Y1158 + Y1162 + Y1163) from human and mouse samples. PA1-26725 is expected to cross react with rat (100% conserved) due to sequence homology.PA1-26725 has been successfully used in Western blot procedures.The PA1-26725 immunogen is a synthetic phosphopeptide corresponding to a portion of IR/IGF1R containing tyrosines 1158, 1162, and 1163 of human insulin receptor (tyrosines 1131, 1135 and 1136 in IGF1R).
Specifications

Specifications

Phospho-IGF1 Rec pTyr1158+1162+1163
Polyclonal
PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide; pH 7.3
P08069, Q60751
IGF1R
Synthetic phosphopeptide corresponding to a portion of IR/IGF1R containing tyrosines 1158, 1162, and 1163 of human insulin receptor.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine
Western Blot
Unconjugated
IGF1R
Purified
Rabbit
IgG
50μL
Store at -20°C
Primary
3480
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit