Learn More
Phospho-IKK-epsilon (Ser172) Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061340
Catalog No. 06-134-0MI
Add to cart
Description
Description
This Anti-phospho-IKK-epsilon (Ser172) Antibody is validated for use in western blotting for the detection of phospho-IKK-epsilon (Ser172).
Specifications
Specifications
phospho-IKK-epsilon (Ser172)
Unconjugated
Affinity Purified
Rabbit
Affinity purified
−20°C for up to 12 months
Polyclonal
Human, Murine, Rat
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide and 30% glycerol.
IKBKE; IKKE; IKKI
Linear peptide corresponding to human IKKepsilon phosphorylated at Ser172.
100μL
Primary
NP_001180251