Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-PDX1 (Thr11) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513046

Catalog No. PIPA513046


Add to cart

Description

Description

This antibody is predicted to react with mouse and rat based on sequence homology.

PDX1 is a transcriptional activator of several genes, including insulin, somatostatin, glucokinase, islet amyloid polypeptide, and glucose transporter type 2. The protein is involved in the early development of the pancreas and plays a major role in glucose-dependent regulation of insulin gene expression. Defects in its gene are a cause of pancreatic agenesis, which can lead to early-onset insulin-dependent diabetes mellitus (NIDDM), as well as maturity onset diabetes of the young type 4 (MODY4).
Specifications

Specifications

Phospho-PDX1 (Thr11)
Polyclonal
PBS with 0.09% sodium azide
P52945
Pancreas/duodenum homeobox protein 1; Insulin promoter factor 1; Islet/duodenum homeobox-1; Somatostatin-transactivating factor 1; Insulin upstream factor 1; Glucose-sensitive factor
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
3651
Dot Blot
Unconjugated
PDX1
Liquid
PDX1
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding T11 of human IPF
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit