Learn More
Phospho-IRS1 (Ser307 mouse/ Ser312 human) Mouse anti-Human, Mouse, Rat, Monkey, Bovine, Canine, Pig, Clone: 24.6.2, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma 051087
Description
This Anti-phospho-IRS1 (Ser307 mouse/ Ser312 human) Antibody, clone 24.6.2 is validated for use in western blot, immunoprecipitation, immunocytochemistry, immunofluorescence for the detection of phospho-IRS1 (Ser307 mouse/ Ser312 human).Specifications
phospho-IRS1 (Ser307 mouse/ Ser312 human) | |
Monoclonal | |
Unconjugated | |
NP_005535.1 | |
Mouse | |
Protein G purfied | |
Primary | |
2-8°C for one year | |
IgG2a κ |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
24.6.2 | |
Purified mouse monoclonal antibody in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide. | |
HIRS-1 , IRS-1 | |
Synthetic peptide corresponding to amino acids surrounding phosphorylated Ser307 of mouse IRS1. | |
100 μg | |
Bovine, Canine, Human, Monkey, Mouse, Porcine, Rat | |