Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-phospho-Jak2 (Tyr1007/Tyr1008), Clone: E132, ™ Non-distribution product as customer accommodation.

Manufacturer:  MilliporeSigma™ 041098/DEL

Catalog No. 50-171-214

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

JAK2 (Janus Kinase 2) belongs to the emerging family of non-receptor Janus tyrosine kinases, which regulate a spectrum of cellular functions downstream of activated cytokine receptors in the lympho-hematopoietic system. Immunological stimuli, such as interferons and cytokines, induce recruitment of Stat transcription factors to cytokine receptor-associated JAK2. JAK2 then phosphorylates proximal Stat factors, which subsequently dimerize, translocate to the nucleus and bind to CIS elements upstream of target gene promoters to regulate transcription. The canonical JAK/Stat pathway is integral to maintaining a normal immune system by stimulating proliferation, differentiation, survival and host resistance to pathogens. Altering JAK/Stat signaling to reduce cytokine induced pro-inflammatory responses represents an attractive target for anti-inflammatory therapies. Within the JAK2 kinase domain, there is a region that has considerable sequence homology to the regulatory region of the insulin receptor.

Host Species: Rabbit
Clone: E132
Isotype: IgG
Cross Reactivity: Human, Mouse, Rat
Immunogen: Phospho-synthetic peptide corresponding to residues surrounding Tyr1007 and Tyr1008 of human Jak2.

Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blotting

Specifications

Specifications

phospho-Jak2 (Tyr1007/Tyr1008)
Unconjugated
Rabbit
IgG
Primary
Human, Murine, Rat
E132
Unpurified
Phospho-synthetic peptide corresponding to residues surrounding Tyr1007 and Tyr1008 of human Jak2.
100μL
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit