Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-phospho-JNK (Thr183 and Tyr185), Polyclonal, Promega™

Rabbit Polyclonal Antibody

$577.50 - $842.50

Specifications

Antigen phospho-JNK (Thr183 and Tyr185)
Clone Polyclonal
Format Affinity Purified
Host Species Rabbit
Immunogen Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
PR-V7931
View Documents
Promega Corporation
V7931
40μL
Each for $577.50
Add to cart
 
PRV7932
View Documents
Promega Corporation
V7932
120μL
Each for $842.50
Add to cart
 
Description

Description

Host Species: Rabbit
Isotype: IgG
Species Reactivity: Many
Immunogen: Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme

Immunocytochemistry, Immunohistochemistry, Western Blotting

Specifications

Specifications

phospho-JNK (Thr183 and Tyr185)
Affinity Purified
Dually phosphorylated Thr/Pro/Tyr region (pTPpY) derived from the catalytic core of the active form of JNK kinase, which corresponds to Thr183 and Tyr185 of the mammalian JNK2 enzyme.
Polyclonal
Polyclonal
Rabbit
IgG
Primary
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit