Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-KIDINS220 (Ser918) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA14251

Catalog No. PA14251

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-4251 detects phosphorylated Kidins220 in human samples. This antibody recognizes the phosphoserine at position 918. PA1-4251 has been successfully used in the Western blot procedure. PA1-4251 immunizing peptide corresponds to amino acid residues 907-923 of the human 220 kDa Kidins220 protein.

A novel protein kinase D-interacting substrate of 220 kDa (Kidins220) is the first identified Protein kinase D (PKD), a serine/threonine kinase. Kidins220 is an integral membrane protein that is expressed in brain and neuroendocrine cells. Kidins220 is phsphorylated by PKD at serine 918 in vivo.
Specifications

Specifications

Phospho-KIDINS220 (Ser918)
Polyclonal
Unconjugated
KIDINS220
Liquid
KIDINS220
Synthetic peptide: R(907) Q M Q R T I T R Q M S(p) F D L T K(923)
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9ULH0
ARMS, MGC163482
Rabbit
IgG
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
57498
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit