Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-KLF4 (Ser245) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA513081

Catalog No. PIPA513081

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This antibody is predicted to react with mouse based on sequence homology.

Transcription factor; can act both as activator and as repressor. Binds the 5 ft.-CACCC-3 ft. core sequence. Binds to the promoter region of its own gene and can activate its own transcription. Regulates the expression of key transcription factors during embryonic development. Plays an important role in maintaining embryonic stem cells, and in preventing their differentiation. Required for establishing the barrier function of the skin and for postnatal maturation and maintece of the ocular surface. Involved in the differentiation of epithelial cells and may also function in skeletal and kidney development. Contributes to the down-regulation of p53/TP53 transcription.
Specifications

Specifications

Phospho-KLF4 (Ser245)
Polyclonal
PBS with 0.09% sodium azide
O43474
Kruppel-like factor 4; EZF; GKLF
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
9314
Dot Blot, Immunocytochemistry, Immunofluorescence
Unconjugated
KLF4
Liquid
KLF4
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S245 of human KLF4
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit