Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-LAT (Tyr132), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126640

Catalog No. PIPA126640

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26640 detects LAT (phospho Y132) from human samples. PA1-26640 is expected to cross react with mouse (85% conserved), rat (77% conserved) due to sequence homology.PA1-26640 has been successfully used in Western blot procedures.The PA1-26640 immunogen is a synthetic phosphopeptide corresponding to a portion of human LAT containing tyrosine 132.
Specifications

Specifications

Phospho-LAT pTyr132
Polyclonal
Unconjugated
LAT
Purified
Rabbit
IgG
50μL
Avoid Freeze/Thaw Cycles
Primary
27040
Western Blot
0.35mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
O43561, O54957, O70601
LAT
Synthetic phosphopeptide corresponding to a portion of human LAT containing tyrosine 132.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit