Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-LCK (Ser158), Polyclonal, Invitrogen™

Catalog No. PIPA126701
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126701 50 μL
1 options

Catalog No. PIPA126701

Supplier: Thermo Scientific PA126701

Rabbit Polyclonal Antibody

PA1-26701 detects Lck (phospho S158) from human samples. PA1-26701 is expected to cross react with mouse due to sequence homology.PA1-26701 has been successfully used in Western blot procedures.The PA1-26701 immunogen is a synthetic phosphopeptide corresponding to a portion of human Lck containing serine 158. The sequence is conserved in human and mouse.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-LCK pSer158
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene LCK
Gene Accession No. P06239, P06240
Gene Symbols LCK
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to a portion of human Lck containing serine 158. The sequence is conserved in human and mouse.
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3932
Target Species Human, Mouse
Content And Storage Store at -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.