Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-LCK (Tyr192), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Thermo Scientific PA126702

Catalog No. PIPA126702

Item Discontinued This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.


Description

Description

PA1-26702 detects Lck (phospho Y192) from human samples. PA1-26702 is expected to cross react with chicken (79% conserved) and mouse (100% conserved) due to sequence homology.PA1-26702 has been successfully used in Western blot procedures.The PA1-26702 immunogen is a synthetic phosphopeptide corresponding to a portion of human Lck containing tyrosine 192. The sequence is conserved in human and mouse.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospho-LCK pTyr192
Polyclonal
Unconjugated
P06239, P06240
Rabbit
Antigen affinity chromatography
RUO
3932
Store at -20°C, Avoid Freeze/Thaw Cycles
Purified
Western Blot
0.5mg/mL
LCK
LCK
Synthetic phosphopeptide corresponding to a portion of human Lck containing tyrosine 192. The sequence is conserved in human and mouse.
50 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit