Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-LEO1 (Ser551) Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA512993

Catalog No. PIPA512993


Add to cart

Description

Description

This antibody is predicted to react with mouse, rat, Xenopus and zebrafish based on sequence homology.

The PAF1 complex is a multifunctional complex. The PAF1 complex interacts with POLR2A. It may be involved in both initiation and elongation, histone methylation and RNA processing. Overexpression of LEO1 induces cell growth arrest and premature senescence of fibroblasts.
Specifications

Specifications

Phospho-LEO1 (Ser551)
Polyclonal
PBS with 0.09% sodium azide
Q8WVC0
RDL; RNA polymerase-associated protein LEO1; Replicative senescence down-regulated leo1-like protein
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
123169
Dot Blot
Unconjugated
LEO1
Liquid
LEO1
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S551 of human Leo1
Protein A, Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit