Learn More
Phospho-ERK1/ERK2 (Thr202, Tyr204), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA112736
Catalog No. 01-674-861
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Extracellular-Signal Regulated Kinase/Mitogen-Activated Protein Kinase (ERK/MAPK) is an integral component of cellular signaling during mitogenesis and differentiation of mitotic cells and also is thought to play a key role in learning and memory. The activity of this kinase is regulated by dual phosphorylation at Thr202 and Tyr204.
In Western blot, immunolabeling is blocked by the phosphopeptide used as antigen but not by the corresponding dephosphopeptide; immunolabeling can be completely eliminated by treatment with lambda-phosphatase. A suggested positive control for this product is Jurkat or rat brain lysate.
Specifications
Specifications
Phospho-ERK1/ERK2 (Thr202, Tyr204)
Polyclonal
HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
P27361, P28482, P63085, Q63844, P21708, P63086, P26696
ERK1, ERT2, Extracellular signal regulated kinase 1, Insulin stimulated MAP2 kinase, MAP kinase 1, MAPK1 antibody, Microtubule associated protein 2 ki
Rabbit
IgG
10 Miniblots
-20° C, Avoid Freeze/Thaw Cycles
Primary
116590, 26413, 26417, 398985, 50689, 5594, 5595
Dot Blot, Western Blot
Unconjugated
MAPK1
Liquid
Mapk1, mapk1.S, MAPK3
Synthetic phosphopeptide corresponding to aa residues surrounding phospho-Thr202 and Tyr204 of ERK/MAPK
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Rat, Xenopus laevis