Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-ERK1/ERK2 (Thr202, Tyr204, Tyr187), Polyclonal, Invitrogen™

Catalog No. PIPA126678
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
PIPA126678 50 μL
1 options

Catalog No. PIPA126678

Supplier: Thermo Scientific PA126678

Rabbit Polyclonal Antibody

kinase involved in intracellular signaling; component of Mapk signaling pathway.

PA1-26678 detects ERK1 + ERK2 (phospho T185 + T202) from human samples. PA1-26678 is expected to cross react with a wide range of species due to sequence homology.PA1-26678 has been successfully used in Western blot procedures.The PA1-26678 immunogen is a synthetic phosphopeptide corresponding to a portion of human ERK 1 + 2 containing threonine 202/185 and tyrosine 204/187. This portion is conserved among many species including rat, mouse, cow, frog, snail, nematode, and fruit
TRUSTED_SUSTAINABILITY

Specifications

Antigen Phospho-p44 MAPK + p42 MAPK pThr202 + pTyr204 + pTyr187
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Gene MAPK1
Gene Accession No. P27361, P28482
Gene Alias Mitogen-activated protein kinase 1, MAP kinase 1, MAPK 1, ERT1, Extracellular signal-regulated kinase 2, ERK-2, MAP kinase isoform p42, p42-MAPK, Mitogen-activated protein kinase 2, MAP kinase 2, MAPK 2 , MAPK1, ERK2, PRKM1, PRKM2, Mitogen-activated protein kinase 3, MAP kinase 3, MAPK 3, ERT2, Extracellular signal-regulated kinase 1, ERK-1, Insulin-stimulated MAP2 kinase, MAP kinase isoform p44, p44-MAPK, Microtubule-associated protein 2 kinase, p44-ERK1, MAPK3, ERK1, PRKM3, p44, p42
Gene Symbols MAPK1, MAPK3
Host Species Rabbit
Immunogen Synthetic phosphopeptide corresponding to a portion of human ERK 1 + 2 containing threonine 202/185 and tyrosine 204/187. This portion is conserved am
Purification Method Antigen affinity chromatography
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5594, 5595
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.