Learn More
Phospho-MEK1/MEK2 (Ser221), Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA126646
Catalog No. PIPA126646
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
PA1-26646 detects MEK1 + MEK2 (phospho S222) from human, mouse, and rat samples. PA1-26646 is expected to cross react with chicken, chimpanzee, and Xenopus laevis due to sequence homology.PA1-26646 has been successfully used in Western blot procedures.The PA1-26646 immunogen is a synthetic phosphopeptide corresponding to a portion of human MEK 1 + 2 containing serine 222. The sequence is conserved in many species including mouse, rat, chimp, chicken, frog, and others.
Specifications
Specifications
Phospho-MEK1 + MEK2 pSer221
Polyclonal
Unconjugated
MAP2K1
Purified
Map2k1, map2k1.L, Map2k2
Synthetic phosphopeptide corresponding to a portion of human MEK 1 + 2 containing serine 222. The sequence is conserved in many species including mous
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Non-human Primate, Rat, Xenopus laevis
Immunocytochemistry, Immunofluorescence, Western Blot
0.5mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
P36507, Q02750, P31938, Q63932, P36506, Q01986, Q05116
MAP2K1, Map2k2, ERK activator kinase 1, MAP kinase kinase 1, MAPK/ERK kinase 1, MAPKK 1, MEK 1, dual specificity mitogen-activated protein kinase kinase 1, mitogen-activated kinase 1, Dual specificity mitogen-activated protein kinase kinase 2, MAP kinase kinase 2, MAPKK 2, ERK activator kinase 2, MAPK/ERK kinase 2, MEK2, MKK2, PRKMK2
Rabbit
IgG
50μL
Avoid Freeze/Thaw Cycles
Primary
5604, 5605