Learn More
Phospho-MEK1 (Thr292) Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14630
Catalog No. PA14630
Add to cart
Description
Description
This antibody is predicted to react with bovine, canine, mouse, non-human primate and rat based on 100% sequence homology. This antibody is specific for the ~45 kDa MEK 1 protein phosphorylated at Thr292 in Western blots of human brain extracts.
MEK 1 (MAP Kinase Kinase, also known as MKK) is an integral component of the MAP kinase cascade that regulates cell growth and differentiation (Ahn, 1993;Chong et al., 2003). This pathway also plays a key role in synaptic plasticity in the brain (Adamsand Sweatt, 2002). Activated MEK 1 acts as a dual specificity kinase phosphorylating both a threonine and a tyrosine residue on MAP kinase (Kyriakis et al., 1991; Seger et al., 1991; Crews et al., 1992). Conversely there also appears to be a feedback phosphorylation of MEK 1 by MAP kinase. The sites on MEK 1 that are phosphorylated by MAP kinase are Thr292 and Thr386 (Mansour et al., 1994).
Specifications
Specifications
Phospho-MEK1 (Thr292)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q02750
phospho-MAP Kinase Kinase, MKK (Thr292)
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
5604
Western Blot
Unconjugated
MAP2K1
Liquid
MAP2K1
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr292 of human MAP2K1 conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Human