Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-SEK1 (Ser257, Thr261), Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126650

Catalog No. PIPA126650

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

PA1-26650 detects MEK4 (phospho S257 + T261) from human samples. PA1-26650 is expected to cross react with mouse (100% conserved) and Xenopus laevis (100% conserved) due to sequence homology.PA1-26650 has been successfully used in Western blot procedures.The PA1-26650 immunogen is a synthetic phosphopeptide corresponding to a portion of human MEK 4 containing serine 257 and threonine 261. The sequence is conserved in mouse and frog.
Specifications

Specifications

Phospho-MEK4/MAP2K4 pSer257+Thr261
Polyclonal
Unconjugated
MAP2K4
Purified
Rabbit
IgG
50μL
Avoid Freeze/Thaw Cycles
Primary
6416
Western Blot
0.5mg/mL
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.3
P45985, P47809
MAP2K4, map2k4.L
Synthetic peptide (Human) derived from a region of human MEK 4 that contains serine 257 and threonine 261 The sequence is conserved in mouse and frog
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Murine, Xenopus laevis
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit