Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

anti-Phospho-Met pTyr1230+1234+1235 Clone: 5H27L59, Invitrogen™

Phospho-Met pTyr1230+1234+1235 Monoclonal antibody 700139 from Thermo Scientific™ Pierce™ specifically detects Phospho-Met pTyr1230+1234+1235 in Human samples, and it is validated for WB and ELISA

Manufacturer:  Invitrogen 700139

Encompass

Catalog No. 70-013-9


Add to cart

Description

Description

c-Met is tyrosine kinase receptor composed of a disulfide-linked heterodimer made of 45 kDa alpha-and 145 kDa beta-subunits found in many tissues. It expresses a transmembrane receptor-like protein, and is involved in regulating cellular proliferation, motility, morphogenesis, and apoptosis. c-Met encodes a high-affinity receptor for hepatocyte growth factor. Additionally, c-Met overexpression has been found in a wide variety of cancer types, particularly lung cancer.

700139 detects Phospho-Met pTyr1230+1234+1235 in Human samples. 700139 has been successfully used in WB and ELISA procedures. The immunogen for 700139 is proprietary.

Applications:

ELISA, Western Blotting

Specifications

Specifications

Phospho-Met pTyr1230+1234+1235
Unconjugated
P08581
Rabbit
IgG
Store at 2 to 8°C for up to 1 month. For long term storage, store at -20°C.
Monoclonal
Human
5H27L59
100μg of Protein A antibody in PBS with 0.09% sodium azide;
Purified
proprietary
Protein A
Primary
4233
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit