Learn More
Phospho-MRE11 (Thr636) Rabbit anti-Human, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Manufacturer:  MilliporeSigma 061374
Catalog No. 06-137-4MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Anti-phospho MRE11 (Thr636) Antibody is a rabbit polyclonal antibody for detection of phospho MRE11 (Thr636) also known as MRE11 meiotic recombination 11 homolog A (S. cerevisiae), has been validated in western blotting.
Specifications
Specifications
phospho MRE11 (Thr636)
Unconjugated
Affinity Purified
Rabbit
Affinity purified
2-8°C for 12 months
Polyclonal
Human
Western Blot
Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
MRE11A; MRE11; MRE11B; ATLD; HNGS1
KLH-conjugated linear peptide corresponding to human MRE11 phosphorylated at Thr636.
100μg
Primary
NP_005581