Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Phospho-MYL1 (Ser20) Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA126470

Catalog No. PA126470


Add to cart

Description

Description

PA1-26470 detects Myosin light chain (phospho S20) from human samples. PA1-26470 is expected to cross react with all mammals and birds due to 100% sequence homology. PA1-26470 has been successfully used in Western blot, IF, ICC, IHC (P) and ELISA procedures. The PA1-26470 immunogen is a synthetic phosphopeptide corresponding to residues K(12) K R P Q R A T (pS) N V F A M F D(27) of the human Myosin Light Chain protein. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. For short-term storage (1-2 weeks), product can be stored at 4°C. For long-term storage, aliquot and store product at -20° C or below, avioiding multiple freeze-thaw cycles.

Myosin is a hexameric ATPase cellular motor protein. It is composed of two heavy chains, two nonphosphorylatable alkali light chains, and two phosphorylatable regulatory light chains. This gene encodes a myosin alkali light chain expressed in fast skeletal muscle. Two transcript variants have been identified for this gene.
Specifications

Specifications

Phospho-MYL1 (Ser20)
Polyclonal
Unconjugated
MYL1
Liquid
MYL1
Synthetic phosphopeptide corresponding to residues K(12) K R P Q R A T (pS) N V F A M F D(27) of the human Myosin Light Chain protein.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
P05976, P05977
MLC1F, MLC3F
Rabbit
IgG
50 μg
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Primary
17901, 4632
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit