Learn More
Phospho-MYPT1 (Thr850) Rabbit anti-Chicken, Human, Clone: SA19, MilliporeSigma™
Rabbit Monoclonal Antibody
Manufacturer:  MilliporeSigma 05773
Catalog No. 05-773-MI
Add to cart
Description
Description
Anti-phospho-MYPT1 (Thr850) Antibody, clone SA19 is an antibody against phospho-MYPT1 (Thr850) for use in western blotting.
Specifications
Specifications
phospho-MYPT1 (Thr850)
SA19
NM_002480
Rabbit
IgG
100μL
Primary
PPP1R12A, MYPT1, MGC133042, MBS
Western Blot
Unconjugated
Purified
KLH-conjugated, synthetic peptide containing the sequence ...RS[pT]GV..., in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1.
Protein A purfied
−20°C for 24 months
Monoclonal
Chicken, Human