Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-MYPT1 (Thr850) Rabbit anti-Chicken, Human, Mouse, Rat, Clone: SA19, MilliporeSigma™
Rabbit Monoclonal Antibody
Supplier: MilliporeSigma 04773
Description
Anti-phospho-MYPT1 (Thr850) Antibody, detects level of phospho-MYPT1 (Thr850) and has been published and validated for use in western blotting.Specifications
phospho-MYPT1 (Thr850) | |
Monoclonal | |
Unconjugated | |
NP_002471.1 | |
KLH-conjugated, synthetic peptide containing the sequence ...RS[pT]GV..., in which [pT] corresponds to phosphorylated threonine at amino acid residue 850 of chicken myosin phosphatase target subunit 1 (MYPT1). Corresponds to Thr853 in human MYPT1. | |
Primary | |
Chicken, Human, Mouse, Rat | |
Supernatant |
Western Blot | |
SA19 | |
Cultured supernantant containing 0.05% sodium azide. | |
Rabbit | |
100 μL | |
MBS, MGC133042, MYPT1 | |
−20°C for 12 months. Aliquot to avoid freeze/thaw cycles. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only