Learn More
Phospho-Neurofilament Mouse anti-Human, Rat, Clone: RMO55, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05774
Catalog No. 05-774-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect phospho-Neurofilament using this Anti-phospho-Neurofilament Antibody, medium chain, clone RMO55 validated for use in western blot, immunohistochemistry.
Specifications
Specifications
phospho-Neurofilament
RMO55
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Ascites
Murine
IgG
200μL
Primary
Human, Rat
Immunohistochemistry, Western Blot
Unconjugated
NM_005382
NEFM, NF-M, NFM, NEF3
HPLC purified rat neurofilament isolated from rat spinal cords
Ascites
−20°C for 24 months.
Monoclonal