Learn More
Phospho-Nibrin/Nbs1 (Ser343) Mouse anti-Human, Clone: 2G2E3, MilliporeSigma™
Mouse Monoclonal Antibody
Manufacturer:  MilliporeSigma 05663
Catalog No. 05-663-MI
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Detect phospho-Nibrin/Nbs1 (Ser343) with Anti-phospho-Nibrin/Nbs1 (Ser343) Antibody, clone 2G2E3 (Mouse Monoclonal Antibody), that has been demonstrated to work in immunoprecipitation.
Specifications
Specifications
phospho-Nibrin/Nbs1 (Ser343)
2G2E3
0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Purified
Murine
κ
200μg
Primary
Human
Immunoprecipitation
Unconjugated
NM_002485
NBN, P95, MGC87362, Nibrin, FLJ10155, ATV, AT-V2, NBS, AT-V1, NBS1
Peptide corresponding to amino acids 337-350 of human Nibrin/nbs1
Protein A chromatography
−20°C for 24 months.
Monoclonal