Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Phospho-NIPA (Ser354) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA538782
Description
Essential component of an SCF-type E3 ligase complex, SCF(NIPA), a complex that controls mitotic entry by mediating ubiquitination and subsequent degradation of cyclin B1 (CCNB1). Its cell-cycle-dependent phosphorylation regulates the assembly of the SCF(NIPA) complex, restricting CCNB1 ubiquitination activity to interphase. Its inactivation results in nuclear accumulation of CCNB1 in interphase and premature mitotic entry. May have an antiapoptotic role in NPM-ALK-mediated signaling events.Specifications
Phospho-NIPA (Ser354) | |
Polyclonal | |
Unconjugated | |
ZC3HC1 | |
1110054L24Rik, NIPA, hNIPA, hematopoietic stem/progenitor cell protein 216, nuclear interacting partner of anaplastic lymphoma kinase (ALK), nuclear interaction partner of ALK, nuclear-interacting partner of ALK, nuclear-interacting partner of anaplastic lymphoma kinase, zinc finge | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
232679, 296957, 51530 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q86WB0, Q80YV2 | |
ZC3HC1 | |
A synthetic phosphopeptide derived from human NIPA around the phosphorylation site of Ser354 (T-R-SP-W-D) | |
100 μg | |
Primary | |
Human, Mouse, Non-human Primate, Rat | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction