Learn More
Phospho-NMDAR2B (Tyr1336) Rabbit anti-Rat, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA14633
Catalog No. PA14633
Add to cart
Description
Description
This antibody is predicted to react with human, mouse and non-human primate based on 100% sequence homology. This antibody is specific for ~180 kDa NMDAR NR2B subunit protein phosphorylated at Tyr1336. Immunolabeling can be blocked by lambda-phosphatase treatment.
NMDAR2B is a member of the N-methyl-D-aspartate (NMDA) class of ionotropic glutamate receptors. NMDA receptors (NMDAR) have been shown to be involved in long-term potentiation, an activity-dependent increase in the efficiency of synaptic transmission thought to underlie certain kinds of memory and learning. NMDAR play a role in neuronal development and has been implicated in several disorders of the central nervous system including Alzheimer’s Disease, epilepsy and ischemic neuronal cell death. Overexpression of the NR2B subunit of the receptor has been associated with increases in learning and memory while aged, memory impaired animals have deficiencies in NR2B expression. Tyr1472 on NR2B is phosphorylated and this may lead to the increased expression of the NMDAR at the synapse that plays a role in synaptic plasticity. NMDR2B is a heterotrimer comprised of seven enetically encoded and differentially expressed subunits that include NR1, NR2 and NR3. Diseases associated with the dysfunction of NMDAR2B include epileptic encephalopathy, autosomal domit 6-related mental retardation.
Specifications
Specifications
Phospho-NMDAR2B (Tyr1336)
Polyclonal
0.01M HEPES with 0.15M NaCl, 100μg/mL BSA, 50% glycerol and no preservative; pH 7.5
Q00960
Glutamate Receptor Ionotropic N Methyl D Aspartate 2B, hNR3, N methyl D aspartate receptor subtype 2B, NR2B
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
24410
Immunohistochemistry, Western Blot
Unconjugated
Grin2b
Liquid
GRIN2B
Synthetic phospho-peptide corresponding to amino acid residues surrounding Tyr1336 of rat GRIN2B conjugated to KLH
Affinity chromatography
RUO
Antibody
Polyclonal
Rat