Learn More
Phospho-p38 MAPK (Thr180, Tyr182) Rabbit anti-Human, Mouse, Rat, Polyclonal, Invitrogen
Rabbit Polyclonal Antibody
Manufacturer:  Invitrogen PA19592
Catalog No. 01-674-466
Add to cart
Description
Description
PA1-9592 detects Phospho-p38 MAP Kinase in human, mouse and rat samples. PA1-9592 has been successfully used in Western blot, immunohistochemistry, and immunocytochemistry procedures. By Western blot, PA1-9592 detects a 43 kDa protein representing Phospho-p38 MAP Kinase. PA1-9592 immunogen is a phosphorylated peptide (pT180-G-pY182) based on human p38 MAP Kinase
Extracellular-Signal Regulated Kinase/Mitogen-Activated Protein Kinase (ERK/MAPK) is an integral component of cellular signaling during mitogenesis and differentiation of mitotic cells and also is thought to play a key role in learning and memory.
Specifications
Specifications
Phospho-p38 MAPK (Thr180, Tyr182)
Polyclonal
Unconjugated
MAPK14
Liquid
MAPK11, MAPK14
Phosphorylated peptide (pT180-G-pY182) based on human p38 MAP Kinase
Affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Immunocytochemistry, Immunohistochemistry, Western Blot
0.5 mg/mL
PBS with no preservative
Q15759, P70618, Q9WUI1, P47811, Q16539
ERK1, ERT2, Extracellular signal regulated kinase 1, Insulin stimulated MAP2 kinase, MAP kinase 1, MAPK1 antibody, Microtubule associated protein 2 ki
Rabbit
IgG
50 μL
-20°C
Primary
1432, 19094, 689314, 81649, 26416, 5600